Potassium selenocyanate, 99%
Potassium selenocyanate, 99%
Potassium selenocyanate, 99%
Thermo Scientific Chemicals

Potassium selenocyanate, 99%

CAS: 3425-46-5 | CKNSe | 144.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 201980100
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS3425-46-5
IUPAC Namepotassium cyanoselanide
Molecular FormulaCKNSe
InChI KeyKYEKHFSRAXRJBR-UHFFFAOYSA-M
SMILES[K+].[Se-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only