Hydrazine monohydrochloride, 98+%
Hydrazine monohydrochloride, 98+%
Hydrazine monohydrochloride, 98+%
Thermo Scientific Chemicals

Hydrazine monohydrochloride, 98+%

CAS: 2644-70-4 | H4N2·HCl | 68.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 201990010
Price (USD)
205.65
Special offer
Online exclusive
Ends: 28-Dec-2026
242.00
Save 36.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
205.65
Special offer
Online exclusive
Ends: 28-Dec-2026
242.00
Save 36.35
Each
Chemical Identifiers
CAS2644-70-4
IUPAC Namedihydrogen hydrazine dichloride
Molecular FormulaCl2H6N2
InChI KeyLIAWOTKNAVAKCX-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration Argentometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only