Sodium persulfate, 98+%
Sodium persulfate, 98+%
Sodium persulfate, 98+%
Sodium persulfate, 98+%
Thermo Scientific Chemicals

Sodium persulfate, 98+%

CAS: 7775-27-1 | Na2O8S2 | 238.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 202020010
Price (EUR)
78,80
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
78,80
Each
Chemical Identifiers
CAS7775-27-1
IUPAC Namedisodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaNa2O8S2
InChI KeyCHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Titration Iodometric98.0 to 102.0 %
Water=<0.15 %
Heavy metals (as Pb)=<20 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only