Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Thermo Scientific Chemicals

Ammonium hexafluorophosphate, 99%, pure

CAS: 16941-11-0 | H4F6NP | 163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 202321000
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Chemical Identifiers
CAS16941-11-0
IUPAC Nameammonium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6H4NP
InChI KeyNIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES[NH4+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Elemental Analysis>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only