Calcon™, pure
Calcon™, pure
Calcon™, pure
Thermo Scientific Chemicals

Calcon™, pure

CAS: 2538-85-4 | C20H13N2NaO5S | 416.383 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 202440250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2538-85-4
IUPAC Namesodium (4E)-4-[2-(2-hydroxynaphthalen-1-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Molecular FormulaC20H13N2NaO5S
InChI KeyYJUGDMOGPJJWDG-QPNALZDCSA-M
SMILES[Na+].OC1=CC=C2C=CC=CC2=C1N\N=C1\C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Dark brown to black
Infrared spectrumConforms
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only