Iron(III) nitrate nonahydrate, 99+%, for analysis
Iron(III) nitrate nonahydrate, 99+%, for analysis
Iron(III) nitrate nonahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, 99+%, for analysis

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 202492500
Price (USD)
83.40
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
83.40
Each
Chemical Identifiers
CAS7782-61-8
IUPAC Nameiron(3+) nonahydrate trinitrate
Molecular FormulaFeH18N3O18
InChI KeySZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light purple
Sodium (Na)=<50 ppm
Potassium (K)=<50 ppm
Arsenic (As)=<0.5 ppm
Appearance (Form)Adhering crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only