Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Potassium sodium tartrate tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium sodium tartrate tetrahydrate, 99+%, for analysis

CAS: 6381-59-5 | C4H4KNaO6·4H2O | 282.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 202865000
Price (EUR)
60,80
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
60,80
Each
Chemical Identifiers
CAS6381-59-5
IUPAC Namepotassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular FormulaC4H12KNaO10
InChI KeyVZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILESO.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Iron (Fe)=<10 ppm
Appearance (Color)Semitransparent white
Titration with HClO499.0 to 102.0 %
Chloride (Cl)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only