Sodium thiosulfate, 98.5%, extra pure, anhydrous
Sodium thiosulfate, 98.5%, extra pure, anhydrous
Sodium thiosulfate, 98.5%, extra pure, anhydrous
Thermo Scientific Chemicals

Sodium thiosulfate, 98.5%, extra pure, anhydrous

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 202875000
Price (USD)
102.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
102.00
Each
Chemical Identifiers
CAS7772-98-7
IUPAC Namedisodium sulfanidesulfonate
Molecular FormulaNa2O3S2
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<1500 ppm
Iron (Fe)=<50 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration Iodimetric98.0 to 102.0 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only