5-Amino-4,6-dichloropyrimidine, 97%
5-Amino-4,6-dichloropyrimidine, 97%
5-Amino-4,6-dichloropyrimidine, 97%
Thermo Scientific Chemicals

5-Amino-4,6-dichloropyrimidine, 97%

CAS: 5413-85-4 | C4H3Cl2N3 | 163.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 203250050
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Chemical Identifiers
CAS5413-85-4
IUPAC Name4,6-dichloropyrimidin-5-amine
Molecular FormulaC4H3Cl2N3
InChI KeyNIGDWBHWHVHOAD-UHFFFAOYSA-N
SMILESNC1=C(Cl)N=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow-brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point143°C to 148°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only