Pierce™ Protein A Agarose
Thermo Scientific™

Pierce™ Protein A Agarose

Thermo Scientific Pierce Protein A Agarose is a standard-capacity Protein A beaded agarose resin for use in a variety ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2033425 mL
203335 mL
Catalog number 20334
Price (USD)
1,654.65
Online exclusive
1,970.00
Save 315.35
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
1,654.65
Online exclusive
1,970.00
Save 315.35
Each
Thermo Scientific Pierce Protein A Agarose is a standard-capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.

Features of Protein A Agarose:

• Native Protein A—immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
• Agarose resin—support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
• Inert and stable—superior manufacturing method immobilizes Protein A by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
• Standard capacity—Pierce Protein A Agarose has a normal load of immobilized Protein A, providing a binding capacity of 12 to 19 mg human IgG/mL resin (approx. 6 to 8 mg mouse IgG/mL resin)

Pierce Protein A Agarose consists of purified native Protein A that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity purification procedure.

Protein A is a cell wall component produced by several strains of Staphylococcus aureus. The 46.7kDa-protein consists of a single polypeptide chain that is essentially devoid of carbohydrate. Native Protein A has contains four high-affinity (Ka = 10^8/mol) binding sites that are capable of interacting with the Fc region of IgG-class antibodies from selected mammalian species. IgG-binding function is optimal at pH 8.2, but efficient binding also occurs in neutral and physiological buffers (pH 7.0 to 7.6).

Protein A is a bacteria-derived protein that binds with high affinity and specificity to the Fc portion of antibodies, especially those of the IgG class. Compared to its alternative (Protein G), Protein A provides the highest binding capacity for subclasses of IgG from rabbits, pigs, dogs and cats. Protein A also has higher binding capacity for human IgG, except for IgG3, which it binds weakly. Protein A also binds weakly to mouse IgG1 and is not recommended for most applications with that antibody isotype.

Pierce Protein A Agarose is prepared using Thermo Scientific AminoLink Coupling Chemistry, which provides several advantages compared to traditional methods of ligand immobilization. AminoLink Immobilization results in conjugation between sugar monomers of the agarose beads and native lysine residues on the Protein A via simple amide bonds. Unlike typical cyanogen bromide (CNBr) immobilization, the AminoLink Method does not introduce any novel chemical groups that could cause undesired nonspecific binding and produces a stable, essentially irreversible bond. The result is a high-binding-capacity resin that retains functional immobilized Protein A through multiple rounds of antibody purification.

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Related Products
Pierce™ Protein A Columns, 1 mL
Pierce™ Protein A IgG Purification Kit, 1 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Purification TargetIgG Antibodies
Quantity25 mL
Sufficient For12 to 19 mg Human IgG/mL Resin, 6 to 8mg mouse IgG/mL Resin
Column TypeAgarose Resin, Affinity
FormatBottle
Product LinePierce
Product TypeAgarose
Stationary PhaseProtein A
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C to 8°C. Product is shipped at ambient temperature.