Lithium iodide, 99%, pure, anhydrous
Lithium iodide, 99%, pure, anhydrous
Lithium iodide, 99%, pure, anhydrous
Thermo Scientific Chemicals

Lithium iodide, 99%, pure, anhydrous

CAS: 10377-51-2 | ILi | 133.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 203592500
Price (USD)
532.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
532.00
Each
Chemical Identifiers
CAS10377-51-2
IUPAC Namelithium(1+) iodide
Molecular FormulaILi
InChI KeyHSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES[Li+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Titration Argentometric>=98.5 % (on anhydrous substance)
Water=<1 % (K.F.)
Appearance (Form)Powder or crystals or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only