(+)-Dihydrocarvone, 98%, mixture of isomers
(+)-Dihydrocarvone, 98%, mixture of isomers
(+)-Dihydrocarvone, 98%, mixture of isomers
Thermo Scientific Chemicals

(+)-Dihydrocarvone, 98%, mixture of isomers

CAS: 5524-05-0 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 203650250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5524-05-0
IUPAC Name(2R,5R)-2-methyl-5-(prop-1-en-2-yl)cyclohexan-1-one
Molecular FormulaC10H16O
InChI KeyAZOCECCLWFDTAP-RKDXNWHRSA-N
SMILESC[C@@H]1CC[C@H](CC1=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 % (sum of isomers)
Specific optical rotation(20°C, 589 nm) (neat) > +10°
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only