(-)-Limonene, 96%
(-)-Limonene, 96%
(-)-Limonene, 96%
Thermo Scientific Chemicals

(-)-Limonene, 96%

CAS: 5989-54-8 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 203735000
Price (USD)
59.90
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.90
Each
Chemical Identifiers
CAS5989-54-8
IUPAC Name(4S)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Molecular FormulaC10H16
InChI KeyXMGQYMWWDOXHJM-SNVBAGLBSA-N
SMILESCC(=C)[C@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation(20°C, 589 nm) (neat) =< -60°
Appearance (Form)Liquid
Refractive index1.4690 to 1.4760 (20°C, 589 nm)
GC>=95 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only