4-Nitroquinoline-N-oxide, 98%
4-Nitroquinoline-N-oxide, 98%
4-Nitroquinoline-N-oxide, 98%
Thermo Scientific Chemicals

4-Nitroquinoline-N-oxide, 98%

CAS: 56-57-5 | C9H6N2O3 | 190.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 203790010
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS56-57-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
Appearance (Color)Ochre to brown
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only