AminoLink™ Coupling Resin, 10 mL - FAQs
View additional product information for AminoLink™ Coupling Resin - FAQs (20382, 20381)
5 product FAQs found
Can you provide the shelf-life for AminoLink Coupling Resin?
AminoLink Coupling Resin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Does sodium azide interfere with AminoLink Support coupling?
No, the gel is stored in sodium azide with no loss in activity. Azide is not a primary amine and is rather unreactive.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between AminoLink Coupling Resin and NHS-Activated Agarose?
Both react with primary amines, but AminoLink Coupling Resin is a two-step conjugation. First a Schiff base is formed between the amine and the aldehyde on the resin, which is then reduced to a stable secondary amine with sodium cyanoborohydride, whereas NHS-Activated Agarose utilizes an NHS ester to form an amide bond in a one-step conjugation.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between AminoLink and AminoLink Plus supports?
AminoLink Plus Coupling Resin is activated at a higher level and has higher flow rates than the original AminoLink Coupling Resin, resulting in higher capacity and faster purification. AminoLink Supports can be used to immobilize any molecule with a primary amine.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How do your AminoLink Coupling Resin and AminoLink Plus Coupling Resin work?
AminoLink and AminoLink Plus Supports are activated with aldehyde groups which will react with primary amines to form Schiff bases, which are reduced to stable, non-reversible secondary amines. Coupling efficiency often exceeds 85% with this support.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.