Dicumarol, 99%
Dicumarol, 99%
Dicumarol, 99%
Thermo Scientific Chemicals

Dicumarol, 99%

CAS: 66-76-2 | C19H12O6 | 336.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 204120050
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.70
Save 7.05
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.70
Save 7.05
Each
Chemical Identifiers
CAS66-76-2
IUPAC Name4-hydroxy-3-[(4-hydroxy-2-oxo-2H-chromen-3-yl)methyl]-2H-chromen-2-one
Molecular FormulaC19H12O6
InChI KeyDOBMPNYZJYQDGZ-UHFFFAOYSA-N
SMILESOC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with TBAH>=98.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only