2,6-Di-tert-butylpyridine, 97%
2,6-Di-tert-butylpyridine, 97%
2,6-Di-tert-butylpyridine, 97%
Thermo Scientific Chemicals

2,6-Di-tert-butylpyridine, 97%

CAS: 585-48-8 | C13H21N | 191.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 204640050
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Chemical Identifiers
CAS585-48-8
IUPAC Name2,6-di-tert-butylpyridine
Molecular FormulaC13H21N
InChI KeyUWKQJZCTQGMHKD-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=CC(=N1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4720 to 1.4740 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only