DL-2,3-Diaminopropionic acid monohydrochloride, 99%
DL-2,3-Diaminopropionic acid monohydrochloride, 99%
DL-2,3-Diaminopropionic acid monohydrochloride, 99%
Thermo Scientific Chemicals

DL-2,3-Diaminopropionic acid monohydrochloride, 99%

CAS: 54897-59-5 | C3H8N2O2·HCl | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 204670050
Price (EUR)
102,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
102,00
Each
Chemical Identifiers
CAS54897-59-5
IUPAC Namehydrogen 2,3-diaminopropanoic acid chloride
Molecular FormulaC3H9ClN2O2
InChI KeySKWCZPYWFRTSDD-UHFFFAOYNA-N
SMILES[H+].[Cl-].NCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only