1-Phenyl-1,2-propanedione-2-oxime, 99%
1-Phenyl-1,2-propanedione-2-oxime, 99%
1-Phenyl-1,2-propanedione-2-oxime, 99%
Thermo Scientific Chemicals

1-Phenyl-1,2-propanedione-2-oxime, 99%

CAS: 119-51-7 | C9H9NO2 | 163.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 205020100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS119-51-7
IUPAC Name(2E)-2-(N-hydroxyimino)-1-phenylpropan-1-one
Molecular FormulaC9H9NO2
InChI KeyYPINLRNGSGGJJT-JXMROGBWSA-N
SMILESC\C(=N/O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point111°C to 115°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only