Sodium bromide, 99+%, extra pure, anhydrous
Sodium bromide, 99+%, extra pure, anhydrous
Sodium bromide, 99+%, extra pure, anhydrous
Thermo Scientific Chemicals

Sodium bromide, 99+%, extra pure, anhydrous

CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 205130010
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
173.00
Save 25.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
173.00
Save 25.35
Each
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 % (on dry substance)
Heavy metals (as Pb)=<10 ppm
Loss on drying=<1.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only