Sodium orthovanadate, 99%
Sodium orthovanadate, 99%
Sodium orthovanadate, 99%
Thermo Scientific Chemicals

Sodium orthovanadate, 99%

CAS: 13721-39-6 ∣ Na3O4V ∣ 183.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 205332500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13721-39-6
IUPAC Nametrisodium oxovanadiumtris(olate)
Molecular FormulaNa3O4V
InChI KeyIHIXIJGXTJIKRB-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[O-][V]([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Silicon (Si)=<200 ppm
Appearance (Color)White to off-white
Assay>=98.5 % (as Na3VO4)
Potassium (K)=<150 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only