Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
Thermo Scientific Chemicals

Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
View all
Plastic drum
Catalog number 205840100
Price (USD)
-
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<1 ppm
Appearance (Form)Clear solution
Color scale=<10 APHA
Titration with HCl28.0 to 30.0 % (As NH3)
Residue after evaporation=<0.002 % (105 °C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only