Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis

CAS: 7783-83-7 | H4FeNO8S2·12H2O | 482.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 205880025
Price (USD)
184.65
Special offer
Online exclusive
Ends: 28-Dec-2026
217.00
Save 32.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
184.65
Special offer
Online exclusive
Ends: 28-Dec-2026
217.00
Save 32.35
Each
Chemical Identifiers
CAS7783-83-7
IUPAC Nameiron(3+) ammonium dodecahydrate disulfate
Molecular FormulaFeH28NO20S2
InChI KeyLCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphate (PO4)=<20 ppm
Potassium (K)=<50 ppm
Chloride (Cl)=<5 ppm
Zinc (Zn)=<10 ppm
Titration Iodometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only