1-Heptanesulfonic Acid, Sodium Salt, 98%
1-Heptanesulfonic Acid, Sodium Salt, 98%
1-Heptanesulfonic Acid, Sodium Salt, 98%
Thermo Scientific Chemicals

1-Heptanesulfonic Acid, Sodium Salt, 98%

CAS: 22767-50-6 | C7H15NaO3S | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 206130250
Price (USD)
156.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
156.00
Each
Chemical Identifiers
CAS22767-50-6
IUPAC Namesodium heptane-1-sulfonate
Molecular FormulaC7H15NaO3S
InChI KeyREFMEZARFCPESH-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or flakes
Loss on drying=<9 % (100°C, 3 h) (vacuum)
Solubility(10 % in water) Clear colorless
Titration after Ion exchange>=97.5 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only