4-Methylpyrazole, 97%
4-Methylpyrazole, 97%
4-Methylpyrazole, 97%
Thermo Scientific Chemicals

4-Methylpyrazole, 97%

CAS: 7554-65-6 | C4H6N2 | 82.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 206970250
Price (USD)
1,500.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,500.00
Each
Chemical Identifiers
CAS7554-65-6
IUPAC Name4-methyl-1H-pyrazole
Molecular FormulaC4H6N2
InChI KeyRIKMMFOAQPJVMX-UHFFFAOYSA-N
SMILESCC1=CNN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4920 to 1.4980 (20°C, 589 nm)
Appearance (Form)Liquid after melting
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only