Nonadecanoic acid, 98+%
Nonadecanoic acid, 98+%
Nonadecanoic acid, 98+%
Thermo Scientific Chemicals

Nonadecanoic acid, 98+%

CAS: 646-30-0 | C19H38O2 | 298.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 207190010
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.50
Save 8.85
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.50
Save 8.85
Each
Chemical Identifiers
CAS646-30-0
IUPAC Namenonadecanoic acid
Molecular FormulaC19H38O2
InChI KeyISYWECDDZWTKFF-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening powder or flakes
GC>=98.0 %
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only