Pipsyl chloride, 97%
Pipsyl chloride, 97%
Pipsyl chloride, 97%
Thermo Scientific Chemicals

Pipsyl chloride, 97%

CAS: 98-61-3 | C6H4ClIO2S | 302.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 207290050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS98-61-3
IUPAC Name4-iodobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClIO2S
InChI KeyPOXFXTSTVWDWIR-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point80°C to 84°C
GC>=96.0 %
Appearance (Color)White to beige to orange
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only