alpha-Terpinene, 90%, tech.
alpha-Terpinene, 90%, tech.
alpha-Terpinene, 90%, tech.
Thermo Scientific Chemicals

alpha-Terpinene, 90%, tech.

CAS: 99-86-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 207491000
Price (USD)
31.65
Online exclusive
35.10
Save 3.45
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Online exclusive
35.10
Save 3.45
Each
Chemical Identifiers
CAS99-86-5
IUPAC Name1-methyl-4-(propan-2-yl)cyclohexa-1,3-diene
Molecular FormulaC10H16
InChI KeyYHQGMYUVUMAZJR-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4770 to 1.4790 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only