Thiocarbohydrazide, 98%
Thiocarbohydrazide, 98%
Thiocarbohydrazide, 98%
Thermo Scientific Chemicals

Thiocarbohydrazide, 98%

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number 207530050
Price (USD)
69.10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
69.10
Each
Chemical Identifiers
CAS2231-57-4
IUPAC Name1,3-diaminothiourea
Molecular FormulaCH6N4S
InChI KeyLJTFFORYSFGNCT-UHFFFAOYSA-N
SMILESNNC(=S)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey
Infrared spectrumConforms
Melting point168°C to 172°C (Dec)
Assay>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only