Sodium tungstate dihydrate, 99+%, for analysis
Sodium tungstate dihydrate, 99+%, for analysis
Sodium tungstate dihydrate, 99+%, for analysis
Sodium tungstate dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium tungstate dihydrate, 99+%, for analysis

CAS: 10213-10-2 | Na2O4W·2H2O | 329.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 207620025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10213-10-2
IUPAC Namedisodium dioxotungstenbis(olate) dihydrate
Molecular FormulaH4Na2O6W
InChI KeyWPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<100 ppm
Chloride (Cl)=<50 ppm
Molybdenum (Mo)=<20 ppm
Assay>=99.0 %
Heavy metals (as Pb)=<10 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
RUO – Research Use Only