Zinc acetate dihydrate, 98%, extra pure
Zinc acetate dihydrate, 98%, extra pure
Zinc acetate dihydrate, 98%, extra pure
Thermo Scientific Chemicals

Zinc acetate dihydrate, 98%, extra pure

CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 207640010
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS5970-45-6
IUPAC Namezinc(2+) diacetate dihydrate
Molecular FormulaC4H10O6Zn
InChI KeyBEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILESO.O.[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Lead (Pb)=<50 ppm
Insoluble matter=<0.005 %
Iron (Fe)=<100 ppm
Appearance (Color)Colorless to white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only