Sodium oxalate, 98.5%, extra pure
Sodium oxalate, 98.5%, extra pure
Sodium oxalate, 98.5%, extra pure
Thermo Scientific Chemicals

Sodium oxalate, 98.5%, extra pure

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207720025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS62-76-0
IUPAC Namedisodium oxalate
Molecular FormulaC2Na2O4
InChI KeyZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with KMnO4>=98.0 %
Insoluble matter=<0.2 % (in water)
Appearance (Form)Powder
Loss on drying=<0.1 % (105°C, 2 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only