Potassium pyrosulfate, 98%, for analysis
Potassium pyrosulfate, 98%, for analysis
Potassium pyrosulfate, 98%, for analysis
Thermo Scientific Chemicals

Potassium pyrosulfate, 98%, for analysis

CAS: 7790-62-7 | K2O7S2 | 254.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207910025
Price (USD)
765.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
765.00
Each
Chemical Identifiers
CAS7790-62-7
IUPAC Namedipotassium (sulfonatooxy)sulfonate
Molecular FormulaK2O7S2
InChI KeyKAQHZJVQFBJKCK-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Phosphate (PO4)=<10 ppm
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only