ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
Thermo Scientific™

ART™ Barrier Pipette Tips in Lift-off Lid Rack

Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelUnit Size
2079E1000 μLART 1000ECase of 3200
2069PK200 μLART 200Pack of 960
2065EPK
also known as 2065E
100 μLART 100EPack of 960
2070300 μLART 300Case of 4800
2070PK
also known as 2070
300 μLART 300Pack of 960
2065E100 μLART 100ECase of 4800
2069200 μLART 200Case of 4800
2065100 μLART 100Case of 4800
2065PK
also known as 2065
100 μLART 100Pack of 960
2079EPK
also known as 2079E
1000 μLART 1000EPack of 800
2139F0.5 to 10 μLART 10FCase of 4800
2139FPK
also known as 2139F
0.5 to 10 μLART 10FPack of 960
2149P20 μLART 20Case of 4800
2149EPK
also known as 2149E
10 μLART 10Pack of 960
2149PPK20 μLART 20Pack of 960
2149E10 μLART 10Case of 4800
Catalog number 2079E
Price (USD)
484.40
Online exclusive
692.00
Save 207.60
Case of 3200
Volume (Metric):
1000 μL
Model:
ART 1000E
Unit Size:
Case of 3200
Price (USD)
484.40
Online exclusive
692.00
Save 207.60
Case of 3200
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier pipette tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 1000E, Filtered, Sterile, Lift-off Lid Rack
ModelART 1000E
Volume (Metric) Dispensing1000μL
AutoclavableNot Autoclavable
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleUniversal
Volume (Metric)1000 μL
Unit SizeCase of 3200