ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
Thermo Scientific™

ART 1000 REACH RACKED barrier sterile pipette tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Catalog number 2079PK
Price (USD)
226.10
Online exclusive
238.00
Save 11.90
Pack of 800
Price (USD)
226.10
Online exclusive
238.00
Save 11.90
Pack of 800
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetting for a variety of applications:

Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 1000 REACH, Filtered, Sterile, Rack
Length (English)4.66 in.
Length (Metric)11.85cm
ModelART 1000 REACH
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizePack of 800