Zinc iodide, 98+%, pure
Zinc iodide, 98+%, pure
Zinc iodide, 98+%, pure
Thermo Scientific Chemicals

Zinc iodide, 98+%, pure

CAS: 10139-47-6 | I2Zn | 319.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 208060500
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.00
Save 6.35 (15%)
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.00
Save 6.35 (15%)
Each
Chemical Identifiers
CAS10139-47-6
IUPAC Namezinc(2+) diiodide
Molecular FormulaI2Zn
InChI KeyUAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES[Zn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Hygroscopic crystalline powder or crystals
Appearance (Color)White to yellow to light beige
Titration Argentometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only