Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company
Thermo Scientific Chemicals

Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company

CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 208080025
Price (USD)
2,227.65
Special offer
Online exclusive
Ends: 28-Dec-2026
2,620.00
Save 392.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
2,227.65
Special offer
Online exclusive
Ends: 28-Dec-2026
2,620.00
Save 392.35
Each
Chemical Identifiers
CAS81133-20-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
Appearance (Color)Brown
Particle size8 to 20 mesh
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gas Adsorbent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only