Ascarite™ II, CO2 absorbent, 20-30 mesh, capacity: 40-50%, product of Arthur H. Thomas Company
Thermo Scientific Chemicals

Ascarite™ II, CO2 absorbent, 20-30 mesh, capacity: 40-50%, product of Arthur H. Thomas Company

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage. | CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Catalog number 208095000
Price (EUR)
361,65
Online Exclusive
402,00
Save 40,35 (10%)
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
361,65
Online Exclusive
402,00
Save 40,35 (10%)
Each
Chemical Identifiers
CAS81133-20-2
SpecificationsSpecification SheetSpecification Sheet
Particle size20 to 30 Mesh
Appearance (Form)Granules
Appearance (Color)Beige to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gas Adsorbent
RUO – Research Use Only