Chlorohydroquinone, 90%, Tech.
Chlorohydroquinone, 90%, Tech.
Chlorohydroquinone, 90%, Tech.
Thermo Scientific Chemicals

Chlorohydroquinone, 90%, Tech.

CAS: 615-67-8 | C6H5ClO2 | 144.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 208221000
Price (USD)
109.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
109.00
Each
Chemical Identifiers
CAS615-67-8
IUPAC Name2-chlorobenzene-1,4-diol
Molecular FormulaC6H5ClO2
InChI KeyAJPXTSMULZANCB-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=88 %
Appearance (Color)Grey to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only