Ethylene glycol diethyl ether, 99%
Ethylene glycol diethyl ether, 99%
Ethylene glycol diethyl ether, 99%
Thermo Scientific Chemicals

Ethylene glycol diethyl ether, 99%

CAS: 629-14-1 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 208241000
Price (USD)
87.60
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
87.60
Each
Chemical Identifiers
CAS629-14-1
IUPAC Name1,2-diethoxyethane
Molecular FormulaC6H14O2
InChI KeyLZDKZFUFMNSQCJ-UHFFFAOYSA-N
SMILESCCOCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.1 %
GC>=98.0 %
Infrared spectrumConforms
Refractive index1.3910 to 1.3930 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only