Methyl 4-hydroxyphenylacetate, 99%
Methyl 4-hydroxyphenylacetate, 99%
Methyl 4-hydroxyphenylacetate, 99%
Thermo Scientific Chemicals

Methyl 4-hydroxyphenylacetate, 99%

CAS: 14199-15-6 | C9H10O3 | 166.18 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 208550250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14199-15-6
IUPAC Namemethyl 2-(4-hydroxyphenyl)acetate
Molecular FormulaC9H10O3
InChI KeyXGDZEDRBLVIUMX-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC=C(O)C=C1
さらに表示
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystals
Melting point55°C to 58°C
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only