Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure
Thermo Scientific™

Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure

Perform light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth PPCO economy bottles with closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2087-000260 mL20 mm
2087-000130 mL20 mm
2087-0004125 mL24 mm
2087-0008250 mL24 mm
2087-0016500 mL28 mm
Catalog number 2087-0002
Price (USD)
117.74
Case of 72
Capacity (Metric):
60 mL
Closure Size:
20 mm
Price (USD)
117.74
Case of 72

Perform light-duty, general-purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure. These polypropylene copolymer bottles are lightweight versions of the original Nalgene Lab Quality bottle. Made from high quality laboratory grade plastic materials for reliably low leachables and extractables.

Features:
  • Economical and recyclable; designed for single use
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials
Note:
  • Completely disengage threads or remove cap before autoclaving
Includes:
  • Polypropylene closures
.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity2 oz., 60 mL
Capacity (English)2 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorNatural
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20 mm
Closure TypeScrew Cap
DescriptionNarrow-Mouth Economy Bottle
Diameter (Metric)37 mm
Diameter (Metric) Inner Neck14 mm
DisposableSingle-use
Height (English)3.34 in.
Height (Metric)85 mm
Neck Finish20 mm
No. per Case72
No. per Pack72
Product TypeNarrow Mouth Bottle
ShapeRound
Through the Closure DispensingNo
Capacity (Metric)60 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 72

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.