Succinic acid, disodium salt, 99%, anhydrous
Succinic acid, disodium salt, 99%, anhydrous
Succinic acid, disodium salt, 99%, anhydrous
Thermo Scientific Chemicals

Succinic acid, disodium salt, 99%, anhydrous

CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 208741000
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.70
Save 4.05
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.70
Save 4.05
Each
Chemical Identifiers
CAS150-90-3
IUPAC Namedisodium butanedioate
Molecular FormulaC4H4Na2O4
InChI KeyZDQYSKICYIVCPN-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Loss on drying=<0.3 % (120°C)
Appearance (Color)White
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only