Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Thermo Scientific Chemicals

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 208750010
Price (USD)
127.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
127.00
Each
Chemical Identifiers
CAS50525-27-4
IUPAC Nameλ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
Molecular FormulaC30H36Cl2N6O6Ru
InChI KeyWHELTKFSBJNBMQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Elemental Analysis>=97.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only