Tetrapropylammonium bromide, 98%
Tetrapropylammonium bromide, 98%
Tetrapropylammonium bromide, 98%
Thermo Scientific Chemicals

Tetrapropylammonium bromide, 98%

CAS: 1941-30-6 | C12H28BrN | 266.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 209371000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1941-30-6
IUPAC Nametetrapropylazanium bromide
Molecular FormulaC12H28BrN
InChI KeyBGQMOFGZRJUORO-UHFFFAOYSA-M
SMILES[Br-].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only