Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS
Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS
Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
数量:
25 mL
100 mL
500 mL
パッケージ:
Glass Bottle
製品番号(カタログ番号) 209565000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
さらに表示
Appearance (Color)Clear colorless
D-Enrichment>=99.80 %
TMS0.02 to 0.04 v/v %
Appearance (Form)Liquid
Water=<0.02 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only