N,N'-Disuccinimidyl carbonate, 98%
N,N'-Disuccinimidyl carbonate, 98%
N,N'-Disuccinimidyl carbonate, 98%
Thermo Scientific Chemicals

N,N'-Disuccinimidyl carbonate, 98%

CAS: 74124-79-1 | C9H8N2O7 | 256.16 g/mol
製品番号(カタログ番号)数量パッケージ
20960025025 gGlass bottle
製品番号(カタログ番号) 209600250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS74124-79-1
IUPAC Namebis(2,5-dioxopyrrolidin-1-yl) carbonate
Molecular FormulaC9H8N2O7
InChI KeyPFYXSUNOLOJMDX-UHFFFAOYSA-N
SMILESO=C(ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
さらに表示
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only