Nalgene™ Fluorinated Narrow-Mouth HDPE Bottles with Closure, 4L, 4000mL; Narrow-mouth - FAQs
View additional product information for Nalgene™ Fluorinated Narrow-Mouth HDPE Bottles with Closure - FAQs (2097-0032PK, 2097-0016, 2097-0005, 2097-0010PK, 2097-0032, 2097-0010, 2097-0005PK, 2097-0016PK, 2097-0008, 2097-0008PK)
1 product FAQs found
To what level are your FLPE fluorinated containers processed?
All of our fluorinated products are processed to Level 5.