2-Brometil-1,3-dioxolano, 97 %, Thermo Scientific Chemicals
2-Brometil-1,3-dioxolano, 97 %, Thermo Scientific Chemicals
2-Brometil-1,3-dioxolano, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Brometil-1,3-dioxolano, 97 %, Thermo Scientific Chemicals

CAS: 4360-63-8 | C4H7BrO2 | 167 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 mL
100 mL
Envase:
Frasco de vidrio
Número de catálogo 209841000
Precio (EUR)
209,00
Each
Cantidad:
100 mL
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
209,00
Each
Identificadores químicos
CAS4360-63-8
IUPAC Name2-(bromomethyl)-1,3-dioxolane
Molecular FormulaC4H7BrO2
InChI KeyCKIIJIDEWWXQEA-UHFFFAOYSA-N
SMILESBrCC1OCCO1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4810 to 1.4840 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only