3-Bromo-1,2-propanediol, 97%
3-Bromo-1,2-propanediol, 97%
3-Bromo-1,2-propanediol, 97%
Thermo Scientific Chemicals

3-Bromo-1,2-propanediol, 97%

CAS: 4704-77-2 | C3H7BrO2 | 154.99 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
產品號碼 209850250
價格 (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS4704-77-2
IUPAC Name3-bromopropane-1,2-diol
Molecular FormulaC3H7BrO2
InChI KeySIBFQOUHOCRXDL-UHFFFAOYNA-N
SMILESOCC(O)CBr
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Appearance (Form)Viscous liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Refractive index1.5165 to 1.5195 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only